Learn More
Biolegend PE/Cyanine7 anti-human CD94
Supplier:  Biolegend 305516
Encompass_Preferred
PE/Cyanine7 anti-human CD94 [DX22]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-114-5284