Learn More
Biolegend PE/Cyanine7 anti-human HLA-E
Supplier:  Biolegend 342607
Encompass_Preferred
PE/Cyanine7 anti-human HLA-E [3D12]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-103-0749