Learn More
Biolegend PE/Cyanine7 anti-human/mouse CD49f
Supplier:  Biolegend 313621
Encompass_Preferred
PE/Cyanine7 anti-human/mouse CD49f [GoH3]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Mouse, Baboon, Capuchin Monkey, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Rhesus, Sheep (Ovine), Swine (Pig, Porcine); Apps: FC; Size: 25 tests
Catalog No. 50-604-114