Learn More
Biolegend PE/Cyanine7 anti-human CD137L (4-1BB Ligand)
Supplier:  Biolegend 311512
Encompass_Preferred
PE/Cyanine7 anti-human CD137L (4-1BB Ligand) [5F4]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1799088