Learn More
Biolegend PE/Cyanine7 anti-human CD15 (SSEA-1)
Supplier:  Biolegend 301924
Encompass_Preferred
PE/Cyanine7 anti-human CD15 (SSEA-1) [HI98]; Isotype: Mouse IgM, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. NC1623343