Learn More
Biolegend PE/Cyanine7 anti-human CD163
Supplier:  Biolegend 333614
Encompass_Preferred
PE/Cyanine7 anti-human CD163 [GHI/61]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC0596841