Learn More
Biolegend PE/Cyanine7 anti-human CD309 (VEGFR2)
Supplier:  Biolegend 359911
Encompass_Preferred
PE/Cyanine7 anti-human CD309 (VEGFR2) [7D4-6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1586133