Learn More
Biolegend PE/Cyanine7 anti-human CD44
Supplier:  Biolegend 338816
Encompass_Preferred
PE/Cyanine7 anti-human CD44 [BJ18]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1400303