Learn More
Biolegend PE/Cyanine7 anti-human CD66b
Supplier:  Biolegend 305116
Encompass_Preferred
PE/Cyanine7 anti-human CD66b [G10F5]; Isotype: Mouse IgM, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests
Catalog No. NC1025607