Learn More
Biolegend PE/Cyanine7 anti-human HLA-G
Supplier:  Biolegend 335912
Encompass_Preferred
PE/Cyanine7 anti-human HLA-G [87G]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1514527