Learn More
Biolegend PE/Cyanine7 anti-mouse CD197 (CCR7)
Supplier:  Biolegend 120124
Encompass_Preferred
PE/Cyanine7 anti-mouse CD197 (CCR7) [4B12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1538009