Learn More
Biolegend PE/Cyanine7 anti-mouse CD366 (Tim-3)
Supplier:  Biolegend 134010
Encompass_Preferred
PE/Cyanine7 anti-mouse CD366 (Tim-3) [B8.2C12]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1417548