Learn More
Biolegend PE/Cyanine7 anti-mouse IL-17A
Supplier:  Biolegend 506922
Encompass_Preferred
PE/Cyanine7 anti-mouse IL-17A [TC11-18H10.1]; Isotype: Rat IgG1, κ; Reactivity: mouse; Apps: ICFC; Size: 100 ug
Catalog No. NC1258424