Learn More
Biolegend PE/Cyanine7 anti-mouse CD205 (DEC-205)
Supplier:  Biolegend 138209
Encompass_Preferred
PE/Cyanine7 anti-mouse CD205 (DEC-205) [NLDC-145]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-029