Learn More
Biolegend PE/Cyanine7 anti-mouse CD206 (MMR)
Supplier:  Biolegend 141719
Encompass_Preferred
PE/Cyanine7 anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 25 ug
Catalog No. 50-605-060