Learn More
Biolegend PE/Cyanine7 anti-mouse CD23
Supplier:  Biolegend 101614
Encompass_Preferred
PE/Cyanine7 anti-mouse CD23 [B3B4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-581