Learn More
Biolegend PE/Cyanine7 anti-mouse/rat/human CD27
Supplier:  Biolegend 124216
Encompass_Preferred
PE/Cyanine7 anti-mouse/rat/human CD27 [LG.3A10]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat, Human; Apps: FC; Size: 100 ug
Catalog No. 50-164-169