Learn More
Biolegend PE/Cyanine7 anti-mouse CD335 (NKp46)
Supplier:  Biolegend 137617
Encompass_Preferred
PE/Cyanine7 anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-403-062