Learn More
Biolegend PE/Cyanine7 anti-mouse CD40
Supplier:  Biolegend 124622
Encompass_Preferred
PE/Cyanine7 anti-mouse CD40 [3/23]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-203