Learn More
Biolegend PE/Cyanine7 anti-mouse CD69
Supplier:  Biolegend 104511
Encompass_Preferred
PE/Cyanine7 anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-942