Learn More
Biolegend PE/Cyanine7 anti-mouse CD86
Supplier:  Biolegend 105116
Encompass_Preferred
PE/Cyanine7 anti-mouse CD86 [PO3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-027