Learn More
Biolegend PE/Cyanine7 anti-mouse CD93 (AA4.1, early B lineage)
Supplier:  Biolegend 136505
Encompass_Preferred
PE/Cyanine7 anti-mouse CD93 (AA4.1, early B lineage) [AA4.1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-920