Learn More
Biolegend PE/Cyanine7 anti-mouse IgG1
Supplier:  Biolegend 406613
Encompass_Preferred
PE/Cyanine7 anti-mouse IgG1 [RMG1-1]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-605-447