Learn More
Biolegend PE/Cyanine7 anti-mouse Ly-6A/E (Sca-1)
Supplier:  Biolegend 108113
Encompass_Preferred
PE/Cyanine7 anti-mouse Ly-6A/E (Sca-1) [D7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-218