Learn More
Biolegend PE/Cyanine7 anti-mouse NK-1.1
Supplier:  Biolegend 108714
Encompass_Preferred
PE/Cyanine7 anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC9978056