Learn More
Biolegend PE/Cyanine7 anti-human Perforin
Supplier:  Biolegend 353316
Encompass_Preferred
PE/Cyanine7 anti-human Perforin [B-D48]; Isotype: Mouse IgG1; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. NC1430658