Learn More
Abcam PE/Cy7 Conjugation Kit - Lightning-Link, 600UG
Catalog No. NC1242894
Encompass_Preferred
The Abcam PE/Cy7 Conjugation Kit ( ab102903) offers several standout features: - Rapid Conjugation: Achieve PE/Cy7 labeling in under 4 hours with just 30 seconds of hands-on time. - High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. - Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence- cited in over 15 publications.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1242894 Supplier Abcam Supplier No. AB102903600UG
The Abcam PE/Cy7 Conjugation Kit ( ab102903) offers several standout features: - Rapid Conjugation: Achieve PE/Cy7 labeling in under 4 hours with just 30 seconds of hands-on time. - High Efficiency: Ensures 100% antibody recovery, preserving the integrity and value of your antibodies. - Versatility: Suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence- cited in over 15 publications.
The product is subject to the following: Abcam Restricted Use Statement