Learn More
Biolegend PE/Cyanine7 Goat anti-rat IgG (minimal x-reactivity)
Supplier:  Biolegend 405413
Encompass_Preferred
PE/Cyanine7 Goat anti-rat IgG (minimal x-reactivity) [Poly4054]; Isotype: Goat Polyclonal IgG; Reactivity: Rat; Apps: FC; Size: 100 ug
Catalog No. 50-169-110