Learn More
Biolegend PE/Cyanine7 anti-H2A.X Phospho (Ser139)
Supplier:  Biolegend 613420
Encompass_Preferred
PE/Cyanine7 anti-H2A.X Phospho (Ser139) [2F3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. NC1686772