Learn More
Cytek Biosciences Inc PE-CY7 MS CD45.1 (A20) 100UG
Supplier:  Cytek Biosciences Inc 600453U100
Encompass_Preferred
PE-Cyanine7 Anti-Mouse CD45.1 (A20) Isotype: Mouse IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-4873