Learn More
Cytek Biosciences Inc PE-CY7 MS LY-6G (1A8) 100UG
Supplier:  Cytek Biosciences Inc 601276U100
Encompass_Preferred
PE-Cyanine7 Anti-Mouse Ly-6G (1A8) Isotype: Rat IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-4893