Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc PE-Cy7 Mu IgG1 Iso Con Anti Is

Catalog No. 5000086609
Encompass_Preferred

Product Name: PE-Cyanine7 Mouse IgG1 Isotype Control (MOPC-21)
Product Class: Antibody
Type: Isotype Control
Product Name: PE-Cyanine7 Mouse IgG1 Isotype Control (MOPC-21)
Cytek Catalog Number: 60-4714-U100
Marker: Isotype Control
Clonality: Monoclonal
Clone: MOPC-21
Host Species: Mouse
Isotype: IgG1
Conjugation: PE-Cyanine7
Size: 100 μg

Catalog No. 50-000-86609 Supplier Cytek Biosciences Inc Supplier No. 604714U100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: PE-Cyanine7 Mouse IgG1 Isotype Control (MOPC-21)
Product Class: Antibody
Type: Isotype Control
Product Name: PE-Cyanine7 Mouse IgG1 Isotype Control (MOPC-21)
Cytek Catalog Number: 60-4714-U100
Marker: Isotype Control
Clonality: Monoclonal
Clone: MOPC-21
Host Species: Mouse
Isotype: IgG1
Conjugation: PE-Cyanine7
Size: 100 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.