Learn More
Biolegend PE/Cyanine7 Rat IgG2a, κ Isotype Ctrl
Supplier:  Biolegend 400522
Encompass_Preferred
PE/Cyanine7 Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 100 ug
Catalog No. NC9074410