Learn More
Biolegend PE/Cyanine7 anti-mouse CD107a (LAMP-1)
Supplier:  Biolegend 121620
Encompass_Preferred
PE/Cyanine7 anti-mouse CD107a (LAMP-1) [1D4B]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 ug
Catalog No. 50-604-950