Learn More
Biolegend PE/Cyanine5 anti-human CD138 S
Catalog No. 502722167
Encompass_Preferred
PE/Cyanine5 anti-human CD138 (Syndecan-1) [MI15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2167 Supplier Biolegend Supplier No. 356547
PE/Cyanine5 anti-human CD138 (Syndecan-1) [MI15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests