Learn More
Biolegend PE/Cyanine5 anti-human CD159a
Catalog No. 502722066
Encompass_Preferred
PE/Cyanine5 anti-human CD159a (NKG2A) [S19004C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2066 Supplier Biolegend Supplier No. 375111
PE/Cyanine5 anti-human CD159a (NKG2A) [S19004C]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests