Learn More
Biolegend PE/Cyanine5 anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Supplier:  Biolegend 344770
Encompass_Preferred
PE/Cyanine5 anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-237-8408