Learn More
Biolegend PE/Cyanine5 anti-mouse CD138 (Syndecan-1)
Supplier:  Biolegend 142539
Encompass_Preferred
PE/Cyanine5 anti-mouse CD138 (Syndecan-1) [281-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-213-1798