Learn More
Biolegend PE/Cyanine5 anti-rat CD161
Supplier:  Biolegend 205612
Encompass_Preferred
PE/Cyanine5 anti-rat CD161 [3.2.3]; Isotype: Mouse IgG1, κ; Reactivity: Rat; Apps: FC; Size: 100 ug
Catalog No. 50-213-1844