Learn More
Biolegend PE/Cyanine7 anti-Bcl-2
Supplier:  Biolegend 633511
Encompass_Preferred
PE/Cyanine7 anti-Bcl-2 [BCL/10C4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Rat; Apps: ICFC; Size: 25 ug
Catalog No. 50-208-6860