Learn More
Biolegend PE/Cyanine7 anti-human CD123
Supplier:  Biolegend 983702
Encompass_Preferred
PE/Cyanine7 anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-207-5573