Learn More
Biolegend PE/Cyanine7 anti-human CD36L1 (SCARB1, SR-BI)
Supplier:  Biolegend 363210
Encompass_Preferred
PE/Cyanine7 anti-human CD36L1 (SCARB1, SR-BI) [m1B9]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8138