Learn More
Biolegend PE/Cyanine7 anti-human CD50 (ICAM-3)
Supplier:  Biolegend 330016
Encompass_Preferred
PE/Cyanine7 anti-human CD50 (ICAM-3) [CBR-IC3/1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-207-5934