Learn More
Biolegend PE/Cyanine7 anti-human/mouse integrin β7
Supplier:  Biolegend 321241
Encompass_Preferred
PE/Cyanine7 anti-human/mouse integrin β7 [FIB504]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Mouse, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-3877