Learn More
Biolegend PE/Cyanine7 anti-human CD158 K
Supplier:  Biolegend 339512
Encompass_Preferred
PE/Cyanine7 anti-human CD158 (KIR2DL1/S1/S3/S5) [HP-MA4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2270