Learn More
Biolegend PE/Cyanine7 anti-human CD218a (IL-18Rα)
Supplier:  Biolegend 313812
Encompass_Preferred
PE/Cyanine7 anti-human CD218a (IL-18Rα) [H44]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1823431