Learn More
Biolegend PE/Cyanine7 anti-human CD235a
Catalog No. 503108068
Encompass_Preferred
PE/Cyanine7 anti-human CD235a (Glycophorin A) Recombinant [QA19A08] Isotype Mouse IgG2a Reactivity Human Apps FC Size 100 tests
Catalog No. 50-310-8068 Supplier Biolegend Supplier No. 379012
PE/Cyanine7 anti-human CD235a (Glycophorin A) Recombinant [QA19A08] Isotype Mouse IgG2a Reactivity Human Apps FC Size 100 tests