Learn More
Biolegend PE/Cyanine7 anti-human CD27
Catalog No. 502721561
Encompass_Preferred
PE/Cyanine7 anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 μL
Catalog No. 50-272-1561 Supplier Biolegend Supplier No. 986902
PE/Cyanine7 anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 μL