Learn More
Biolegend PE/Cyanine7 anti-human CD29
Catalog No. 502722489
Encompass_Preferred
PE/Cyanine7 anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2489 Supplier Biolegend Supplier No. 303025
PE/Cyanine7 anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests