Learn More
Biolegend PE/Cyanine7 anti-human CD32B/C [S18005H]; Isotype: Mouse IgG1, ; Reactivity: Human; Apps: FC; Size: 25 tests
Supplier:  Biolegend 398313
Encompass_Preferred
PE/Cyanine7 anti-human CD32B/C [S18005H]; Isotype: Mouse IgG1, ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9367